Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR7A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012067.2 | NP_036199.2 | 5 | 696 | ||
XM_011541046.2 | XP_011539348.1 | 5 | 1272 | |||
XM_017000714.1 | XP_016856203.1 | 5 | 430 | |||
GenBank mRNA | AF040639.1 | - | 5 | 703 | ||
AJ271799.1 | - | 5 | 626 | |||
BC025709.1 | - | 5 | 682 | 5224626 | ||
BC031562.1 | - | 5 | 638 | 5184829 | ||
BC042420.1 | - | 5 | 662 | 5186889 | ||
BM920315.1 | - | 5 | 660 |
Gene Symbol: AKR7L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040288.1 | 5 | 619 | |||
GenBank mRNA | AJ278012.1 | - | 4 | 405 |