Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164814.1 | NP_001158286.1 | 6 | 1117 | ||
NM_001164815.1 | NP_001158287.1 | 5 | 997 | |||
NM_014977.3 | NP_055792.1 | 6 | 1117 | |||
XM_005267415.3 | XP_005267472.1 | 6 | 921 | |||
XM_005267416.3 | XP_005267473.1 | 6 | 921 | |||
XM_006720081.3 | XP_006720144.1 | 6 | 921 | |||
XM_011536570.2 | XP_011534872.1 | 6 | 921 | |||
GenBank mRNA | AB014570.2 | - | 6 | 626 | ||
AF124726.1 | - | 6 | 1116 | |||
AK299034.1 | - | 5 | 502 | |||
BC140805.1 | - | 6 | 886 | 9021673 |