Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270520.1 | NP_001257449.1 | 21 | 2729 | ||
NM_014992.2 | NP_055807.1 | 21 | 2653 | |||
XM_005267430.1 | XP_005267487.1 | 22 | 2811 | |||
XM_005267431.1 | XP_005267488.1 | 22 | 2911 | |||
GenBank mRNA | AB014566.1 | - | 21 | 2639 | ||
AK021890.1 | - | 11 | 1077 | |||
AK093813.1 | - | 5 | 410 | |||
AK291790.1 | - | 21 | 2698 | |||
BC024781.1 | - | 11 | 1370 | |||
BC038428.1 | - | 21 | 2721 | 4993504 | ||
BC064999.1 | - | 21 | 2712 | 5784628 |