Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHLPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289003.1 | NP_001275932.1 | 12 | 2451 | ||
NM_015020.3 | NP_055835.2 | 12 | 2104 | |||
GenBank mRNA | AB023148.2 | - | 12 | 2451 | ||
BC035267.1 | - | 7 | 939 | |||
BC129927.1 | - | 11 | 1731 | 40124720 | ||
BX647823.1 | - | 12 | 2105 | |||
BX648157.1 | - | 6 | 840 |