Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPO7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015024.4 | NP_055839.3 | 27 | 3240 | ||
XM_017013244.1 | XP_016868733.1 | 27 | 3494 | |||
XM_017013245.1 | XP_016868734.1 | 27 | 3261 | |||
XM_017013246.1 | XP_016868735.1 | 27 | 3463 | |||
GenBank mRNA | AB018288.1 | - | 24 | 2604 | ||
AF064729.1 | - | 27 | 3150 | |||
AK291472.1 | - | 27 | 3155 | |||
AK299848.1 | - | 23 | 2676 | |||
BC014219.2 | - | 4 | 499 | |||
BC030785.1 | - | 27 | 3239 | 5269230 |