Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPO7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015024.4 | NP_055839.3 | 9 | 974 | ||
XM_017013244.1 | XP_016868733.1 | 9 | 1228 | |||
XM_017013245.1 | XP_016868734.1 | 9 | 995 | |||
XM_017013246.1 | XP_016868735.1 | 9 | 1197 | |||
XM_017013247.1 | XP_016868736.1 | 9 | 1388 | |||
GenBank mRNA | AF064729.1 | - | 9 | 884 | ||
AK291472.1 | - | 9 | 889 | |||
BC030785.1 | - | 9 | 973 | 5269230 |