Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WAPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318328.1 | NP_001305257.1 | 10 | 2836 | ||
NM_015045.3 | NP_055860.1 | 10 | 2854 | |||
XM_011539548.1 | XP_011537850.1 | 10 | 2904 | |||
XM_017015979.1 | XP_016871468.1 | 9 | 2751 | |||
XM_017015980.1 | XP_016871469.1 | 9 | 2694 | |||
XM_017015981.1 | XP_016871470.1 | 7 | 2441 | |||
GenBank mRNA | AB065003.1 | - | 10 | 2857 | ||
AF479418.1 | - | 11 | 2532 | |||
AK027005.1 | - | 3 | 577 | |||
BC017393.1 | - | 3 | 290 | |||
BC140858.1 | - | 10 | 2560 | 9021726 | ||
BC150269.1 | - | 11 | 2705 | 9007232 | ||
D87450.1 | - | 11 | 2705 |