Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF423 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271620.2 | NP_001258549.1 | 4 | 3161 | ||
NM_015069.4 | NP_055884.2 | 4 | 3051 | |||
XM_005255856.4 | XP_005255913.1 | 4 | 3764 | |||
XM_005255857.4 | XP_005255914.1 | 2 | 3254 | |||
XM_006721171.3 | XP_006721234.1 | 7 | 3493 | |||
XM_017023076.1 | XP_016878565.1 | 4 | 3778 | |||
XM_017023077.1 | XP_016878566.1 | 4 | 3127 | |||
XM_017023078.1 | XP_016878567.1 | 3 | 3100 | |||
GenBank mRNA | AB018303.2 | - | 2 | 2818 | ||
AF221712.1 | - | 2 | 2818 | |||
AK027479.1 | - | 1 | 1217 | |||
AK304829.1 | - | 2 | 2855 | |||
AK309835.1 | - | 2 | 2836 | |||
AK314742.1 | - | 4 | 3162 | |||
BC112315.1 | - | 4 | 3053 | 8327783 | ||
BC112317.1 | - | 4 | 3053 | 8327785 |