Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135608.1 | NP_001129080.1 | 15 | 1360 | ||
NM_015071.4 | NP_055886.1 | 15 | 1360 | |||
XM_005268398.4 | XP_005268455.1 | 15 | 1597 | |||
XM_005268399.4 | XP_005268456.1 | 15 | 1594 | |||
XM_005268400.4 | XP_005268457.1 | 15 | 1595 | |||
XM_005268402.4 | XP_005268459.1 | 15 | 1714 | |||
XM_006714774.3 | XP_006714837.1 | 15 | 1601 | |||
XM_011537610.2 | XP_011535912.1 | 15 | 1600 | |||
XM_011537611.2 | XP_011535913.1 | 15 | 1600 | |||
XM_017009247.1 | XP_016864736.1 | 15 | 1600 | |||
XM_017009248.1 | XP_016864737.1 | 15 | 1596 | |||
XM_017009249.1 | XP_016864738.1 | 15 | 1596 | |||
XM_017009250.1 | XP_016864739.1 | 15 | 1596 | |||
GenBank mRNA | AB014521.1 | - | 14 | 1142 | ||
AK095220.1 | - | 5 | 473 | |||
BC021059.2 | - | 1 | 40 | |||
BC068555.1 | - | 15 | 1680 | 30343863 | ||
KU178561.1 | - | 15 | 1325 | |||
KU178562.1 | - | 15 | 1325 |