Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135608.1 | NP_001129080.1 | 16 | 1432 | ||
NM_015071.4 | NP_055886.1 | 16 | 1432 | |||
XM_005268398.4 | XP_005268455.1 | 16 | 1669 | |||
XM_005268399.4 | XP_005268456.1 | 16 | 1666 | |||
XM_005268400.4 | XP_005268457.1 | 16 | 1667 | |||
XM_005268402.4 | XP_005268459.1 | 16 | 1786 | |||
XM_006714774.3 | XP_006714837.1 | 16 | 1673 | |||
XM_011537610.2 | XP_011535912.1 | 16 | 1672 | |||
XM_011537611.2 | XP_011535913.1 | 16 | 1672 | |||
XM_017009247.1 | XP_016864736.1 | 16 | 1672 | |||
XM_017009248.1 | XP_016864737.1 | 16 | 1668 | |||
XM_017009249.1 | XP_016864738.1 | 16 | 1668 | |||
XM_017009250.1 | XP_016864739.1 | 16 | 1668 | |||
GenBank mRNA | AB014521.1 | - | 15 | 1214 | ||
AK095220.1 | - | 6 | 545 | |||
BC021059.2 | - | 2 | 112 | |||
BC068555.1 | - | 16 | 1752 | 30343863 | ||
KU178561.1 | - | 16 | 1397 | |||
KU178562.1 | - | 16 | 1397 |