Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135608.1 | NP_001129080.1 | 19 | 1857 | ||
NM_015071.4 | NP_055886.1 | 19 | 1857 | |||
XM_005268398.4 | XP_005268455.1 | 19 | 2094 | |||
XM_005268399.4 | XP_005268456.1 | 19 | 2091 | |||
XM_005268400.4 | XP_005268457.1 | 19 | 2092 | |||
XM_005268402.4 | XP_005268459.1 | 19 | 2211 | |||
XM_017009247.1 | XP_016864736.1 | 19 | 2097 | |||
XM_017009248.1 | XP_016864737.1 | 19 | 2093 | |||
XM_017009249.1 | XP_016864738.1 | 19 | 2093 | |||
XM_017009250.1 | XP_016864739.1 | 19 | 2093 | |||
GenBank mRNA | AB014521.1 | - | 18 | 1639 | ||
AK095220.1 | - | 9 | 970 | |||
AK298708.1 | - | 1 | 122 | |||
BC021059.2 | - | 5 | 537 | |||
BC068555.1 | - | 19 | 2177 | 30343863 | ||
HM037040.1 | - | 2 | 191 | |||
KU178561.1 | - | 19 | 1822 | |||
KU178562.1 | - | 19 | 1822 |