Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015072.4 | NP_055887.3 | 14 | 1411 | ||
GenBank mRNA | AB023215.1 | - | 12 | 971 | ||
AK309748.1 | - | 14 | 1351 | |||
AY237126.1 | - | 14 | 1341 | |||
BC136472.1 | - | 14 | 1301 | 9020461 | ||
BC136473.1 | - | 14 | 1301 | 9020462 | ||
BX647530.1 | - | 7 | 442 |