Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015072.4 | NP_055887.3 | 26 | 3063 | ||
GenBank mRNA | AB023215.1 | - | 24 | 2623 | ||
AK309748.1 | - | 27 | 3045 | |||
AL136808.1 | - | 12 | 1511 | |||
AY237126.1 | - | 26 | 2993 | |||
BC016625.1 | - | 13 | 1644 | |||
BC136472.1 | - | 26 | 2953 | 9020461 | ||
BC136473.1 | - | 26 | 2953 | 9020462 | ||
BX647530.1 | - | 19 | 2132 | |||
CR533554.1 | - | 11 | 1441 |