Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015074.3 | NP_055889.2 | 17 | 1844 | ||
NM_183416.3 | NP_904325.2 | 17 | 1844 | |||
GenBank mRNA | AB011163.2 | - | 19 | 1931 | ||
AB017133.1 | - | 18 | 1814 | |||
AB040881.2 | - | 17 | 1903 | |||
AB088210.1 | - | 18 | 1730 | |||
AB088211.1 | - | 17 | 1688 | |||
AB088212.1 | - | 18 | 1652 | |||
AB088213.1 | - | 17 | 1610 | |||
AF257176.1 | - | 17 | 1745 | |||
AY043362.1 | - | 16 | 1596 | |||
AY139835.1 | - | 17 | 1745 | |||
BC115395.2 | - | 16 | 1680 | 40067993 |