Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135776.1 | NP_001129248.1 | 2 | 695 | ||
NM_014007.3 | NP_054726.1 | 3 | 818 | |||
XM_005251835.3 | XP_005251892.1 | 3 | 1139 | |||
XM_011518409.1 | XP_011516711.1 | 4 | 831 | |||
XM_011518411.2 | XP_011516713.1 | 2 | 945 | |||
GenBank mRNA | AB007874.1 | - | 1 | 1677 | ||
AF049907.1 | - | 3 | 798 | |||
AK026123.1 | - | 3 | 764 | |||
BC008828.2 | - | 3 | 742 | 4123095 | ||
BT007194.1 | - | 1 | 545 |