Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015079.5 | NP_055894.6 | 3 | 634 | ||
NM_144572.1 | NP_653173.1 | 3 | 634 | |||
XM_011521387.2 | XP_011519689.1 | 3 | 690 | |||
XM_017022022.1 | XP_016877511.1 | 3 | 510 | |||
GenBank mRNA | AB028978.1 | - | 3 | 497 | ||
BC140766.1 | - | 2 | 279 | 9021634 | ||
BC144482.1 | - | 2 | 279 | 9053009 | ||
BC152465.1 | - | 3 | 497 | 8862601 |