Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPG20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142294.1 | NP_001135766.1 | 8 | 1762 | ||
NM_001142295.1 | NP_001135767.1 | 8 | 1886 | |||
NM_001142296.1 | NP_001135768.1 | 9 | 1938 | |||
NM_015087.4 | NP_055902.1 | 8 | 1955 | |||
XM_005266313.4 | XP_005266370.1 | 8 | 1880 | |||
XM_005266314.3 | XP_005266371.1 | 9 | 1929 | |||
XM_005266315.3 | XP_005266372.1 | 9 | 1783 | |||
XM_005266317.3 | XP_005266374.1 | 8 | 1713 | |||
XM_011535012.2 | XP_011533314.1 | 8 | 1866 | |||
GenBank mRNA | AB011182.1 | - | 8 | 1729 | ||
AK002207.1 | - | 7 | 906 | |||
AK057764.1 | - | 9 | 1275 | |||
AK291724.1 | - | 8 | 1806 | |||
AK302119.1 | - | 7 | 947 | |||
AY038359.1 | - | 7 | 1668 | |||
AY123329.1 | - | 8 | 1729 | |||
BC047083.1 | - | 8 | 1762 | 5313378 | ||
BX648004.1 | - | 8 | 1851 | |||
KU178563.1 | - | 7 | 1668 |