Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPG20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142294.1 | NP_001135766.1 | 8, 9 | 1835 | ||
NM_001142295.1 | NP_001135767.1 | 8, 9 | 1959 | |||
NM_001142296.1 | NP_001135768.1 | 9, 10 | 2011 | |||
NM_015087.4 | NP_055902.1 | 8, 9 | 2028 | |||
XM_005266313.4 | XP_005266370.1 | 8, 9 | 1953 | |||
XM_005266314.3 | XP_005266371.1 | 9, 10 | 2002 | |||
XM_005266315.3 | XP_005266372.1 | 9, 10 | 1856 | |||
XM_005266317.3 | XP_005266374.1 | 8, 9 | 1786 | |||
XM_011535012.2 | XP_011533314.1 | 8, 9 | 1939 | |||
GenBank mRNA | AB011182.1 | - | 8, 9 | 1802 | ||
AK002207.1 | - | 7, 8 | 979 | |||
AK291724.1 | - | 8, 9 | 1879 | |||
AK302119.1 | - | 7, 8 | 1020 | |||
AY038359.1 | - | 7, 8 | 1741 | |||
AY123329.1 | - | 8, 9 | 1802 | |||
BC047083.1 | - | 8, 9 | 1835 | 5313378 | ||
BX648004.1 | - | 8, 9 | 1924 | |||
KU178563.1 | - | 7, 8 | 1741 |