Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COLGALT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303420.1 | NP_001290349.1 | 10 | 1767 | ||
NM_001303421.1 | NP_001290350.1 | 10 | 1281 | |||
NM_015101.3 | NP_055916.1 | 10 | 1767 | |||
XM_011509332.2 | XP_011507634.1 | 11 | 1573 | |||
XM_011509334.2 | XP_011507636.1 | 11 | 1380 | |||
XM_011509335.2 | XP_011507637.1 | 11 | 1431 | |||
XM_011509336.2 | XP_011507638.1 | 11 | 1208 | |||
XM_011509337.2 | XP_011507639.1 | 7 | 1507 | |||
XM_011509338.2 | XP_011507640.1 | 8 | 1259 | |||
XM_017000750.1 | XP_016856239.1 | 10 | 1332 | |||
XM_017000751.1 | XP_016856240.1 | 5 | 1191 | |||
GenBank mRNA | AB011156.1 | - | 10 | 1692 | ||
AF288389.1 | - | 10 | 1726 | |||
AK095090.1 | - | 5 | 1002 | |||
AK095192.1 | - | 10 | 1255 | |||
AK293973.1 | - | 10 | 1474 | |||
BC035672.1 | - | 10 | 1749 | 5740842 |