Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319245.1 | NP_001306174.1 | 10 | 1286 | ||
NM_001324320.1 | NP_001311249.1 | 11 | 1313 | |||
NM_001324321.1 | NP_001311250.1 | 10 | 1399 | |||
NM_015112.2 | NP_055927.2 | 10 | 1286 | |||
XM_005270655.3 | XP_005270712.1 | 9 | 1006 | |||
XM_005270656.4 | XP_005270713.1 | 9 | 875 | |||
XM_006710477.3 | XP_006710540.1 | 8 | 683 | |||
XM_011541059.2 | XP_011539361.1 | 12 | 1520 | |||
XM_011541061.2 | XP_011539363.1 | 12 | 1520 | |||
XM_011541062.2 | XP_011539364.1 | 11 | 1499 | |||
XM_011541063.1 | XP_011539365.1 | 12 | 1084 | |||
XM_011541064.2 | XP_011539366.1 | 12 | 1081 | |||
XM_011541065.1 | XP_011539367.1 | 12 | 1183 | |||
XM_011541067.2 | XP_011539369.1 | 8 | 997 | |||
XM_011541068.1 | XP_011539370.1 | 7 | 614 | |||
XM_011541069.2 | XP_011539371.1 | 12 | 1520 | |||
XM_017000752.1 | XP_016856241.1 | 10 | 1513 | |||
XM_017000753.1 | XP_016856242.1 | 9 | 847 | |||
XM_017000754.1 | XP_016856243.1 | 8 | 826 | |||
XM_017000755.1 | XP_016856244.1 | 9 | 1318 | |||
GenBank mRNA | AB047005.1 | - | 10 | 1286 | ||
AK027123.1 | - | 8 | 825 | |||
AK096519.1 | - | 10 | 712 | |||
AK309510.1 | - | 9 | 1008 | |||
BC065499.1 | - | 10 | 1260 | 5499830 |