Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCUN1D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040402.2 | NP_001035492.1 | 11 | 1090 | ||
NM_001287755.1 | NP_001274684.1 | 11 | 986 | |||
NM_001287757.1 | NP_001274686.1 | 12 | 1253 | |||
NM_015115.3 | NP_055930.2 | 10 | 985 | |||
XM_005265731.3 | XP_005265788.1 | 10 | 875 | |||
XM_011534379.2 | XP_011532681.1 | 11 | 1513 | |||
XM_011534380.1 | XP_011532682.1 | 11 | 849 | |||
XM_017007912.1 | XP_016863401.1 | 10 | 838 | |||
XM_017007913.1 | XP_016863402.1 | 10 | 1428 | |||
GenBank mRNA | AK027051.1 | - | 11 | 909 | ||
AK124346.1 | - | 9 | 1553 | |||
AK294894.1 | - | 11 | 986 | |||
AK294896.1 | - | 12 | 1100 | |||
BC053897.1 | - | 10 | 851 | 6173607 | ||
BX537944.1 | - | 11 | 917 | |||
D87466.1 | - | 11 | 900 |