Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015138.4 | NP_055953.3 | 18 | 2051 | ||
GenBank mRNA | AK292302.1 | - | 18 | 2023 | ||
BC015052.2 | - | 18 | 2001 | 3909737 | ||
D87440.2 | - | 18 | 2017 |