Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015150.1 | NP_055965.1 | 6 | 1155 | ||
XM_005264985.1 | XP_005265042.1 | 6 | 1112 | |||
XM_005264986.2 | XP_005265043.1 | 6 | 1009 | |||
XM_006713069.1 | XP_006713132.1 | 6 | 1018 | |||
XM_011533529.1 | XP_011531831.1 | 8 | 1552 | |||
XM_011533530.1 | XP_011531832.1 | 6 | 1180 | |||
XM_011533531.1 | XP_011531833.1 | 8 | 1552 | |||
XM_017006005.1 | XP_016861494.1 | 8 | 1552 | |||
XM_017006006.1 | XP_016861495.1 | 8 | 1552 | |||
XM_017006007.1 | XP_016861496.1 | 6 | 1139 | |||
GenBank mRNA | AK074446.1 | - | 6 | 961 | ||
AK301024.1 | - | 5 | 848 | |||
AK303846.1 | - | 2 | 152 | |||
AY237654.1 | - | 6 | 1146 | |||
BC006400.2 | - | 2 | 83 | |||
BC014032.2 | - | 2 | 83 | |||
BC032349.1 | - | 5 | 828 | |||
BC051336.1 | - | 6 | 932 | |||
BC069209.1 | - | 6 | 932 | |||
BC098270.1 | - | 6 | 979 | 40001191 | ||
BC100667.1 | - | 6 | 979 | 40001188 | ||
BC100669.1 | - | 6 | 979 | 40001190 | ||
BC121795.1 | - | 6 | 979 | 40011545 | ||
D42043.1 | - | 6 | 1083 |
Gene Symbol: OXNAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017007489.1 | XP_016862978.1 | 10 | 2020 |