Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KANK1 SIRNA Assays ›
Gene Symbol:
KANK1
Gene Name:
KN motif and ankyrin repeat domains 1
Gene Aliases:
ANKRD15, CPSQ2, KANK
Chromosome Location:
Chr.9: 470294 - 746106 on Build GRCh38
UniGene ID:
Hs.306764
Species:
Human
siRNA ID
HSS118332
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KANK1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256876.1
NP_001243805.1
7
1520
NM_001256877.1
NP_001243806.1
4
1629
NM_015158.3
NP_055973.2
3
1014
NM_153186.4
NP_694856.1
2
1276
XM_005251411.2
XP_005251468.1
3
1014
XM_005251414.3
XP_005251471.1
2
1477
XM_005251415.2
XP_005251472.1
2
1206
XM_005251416.2
XP_005251473.1
2
982
XM_005251417.3
XP_005251474.1
2
1473
XM_005251418.2
XP_005251475.1
2
1484
XM_005251419.2
XP_005251476.1
2
1175
XM_006716743.2
XP_006716806.1
3
990
XM_011517819.1
XP_011516121.1
4
1105
XM_011517820.1
XP_011516122.1
3
1122
XM_011517821.1
XP_011516123.1
3
1049
XM_017014511.1
XP_016870000.1
4
1227
XM_017014512.1
XP_016870001.1
3
2538
XM_017014513.1
XP_016870002.1
5
1740
XM_017014514.1
XP_016870003.1
3
1426
XM_017014515.1
XP_016870004.1
4
1162
XM_017014516.1
XP_016870005.1
3
1426
XM_017014517.1
XP_016870006.1
3
1426
XM_017014518.1
XP_016870007.1
3
1014
XM_017014519.1
XP_016870008.1
3
1426
XM_017014520.1
XP_016870009.1
3
1014
XM_017014521.1
XP_016870010.1
3
1426
XM_017014522.1
XP_016870011.1
4
1227
XM_017014523.1
XP_016870012.1
3
1014
XM_017014524.1
XP_016870013.1
3
1426
XM_017014525.1
XP_016870014.1
3
1426
XM_017014526.1
XP_016870015.1
3
1459
XM_017014527.1
XP_016870016.1
2
1206
XM_017014528.1
XP_016870017.1
3
1060
XM_017014529.1
XP_016870018.1
2
1473
XM_017014530.1
XP_016870019.1
2
982
XM_017014531.1
XP_016870020.1
2
1206
XM_017014532.1
XP_016870021.1
2
1480
XM_017014533.1
XP_016870022.1
2
923
XM_017014534.1
XP_016870023.1
2
1206
XM_017014535.1
XP_016870024.1
2
1472
XM_017014536.1
XP_016870025.1
2
1175
XM_017014537.1
XP_016870026.1
2
995
XM_017014538.1
XP_016870027.1
2
1480
XM_017014539.1
XP_016870028.1
2
995
XM_017014540.1
XP_016870029.1
2
1471
XM_017014541.1
XP_016870030.1
2
1475
XM_017014542.1
XP_016870031.1
2
982
GenBank mRNA
AK026421.1
-
1
627
AK290028.1
-
3
980
AK292989.1
-
3
993
AL832454.2
-
3
1006
AL833161.1
-
3
994
BC037495.2
-
7
1523
5265517
BC038116.2
-
2
1186
5261060
BX648931.1
-
3
1008
D79994.2
-
3
1579
Back To Top
More Information
Panther Classification:
Molecular Function -
non-motor actin binding protein
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
negative regulation of neuron projection development
positive regulation of Wnt signaling pathway
negative regulation of cell migration
negative regulation of actin filament polymerization
negative regulation of Rho protein signal transduction
positive regulation of catenin import into nucleus
negative regulation of insulin receptor signaling pathway
positive regulation of wound healing
negative regulation of substrate adhesion-dependent cell spreading
negative regulation of ruffle assembly
regulation of establishment of cell polarity
negative regulation of lamellipodium morphogenesis
protein binding
beta-catenin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-69b65fdbbd-sq4fc:80/100.66.76.194:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline