Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GANAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278192.1 | NP_001265121.1 | 8 | 906 | ||
NM_001278193.1 | NP_001265122.1 | 7 | 840 | |||
NM_001278194.1 | NP_001265123.1 | 9 | 1073 | |||
NM_198334.2 | NP_938148.1 | 10 | 1182 | |||
NM_198335.3 | NP_938149.2 | 11 | 1248 | |||
XM_017017412.1 | XP_016872901.1 | 8 | 916 | |||
XM_017017413.1 | XP_016872902.1 | 10 | 1298 | |||
XM_017017414.1 | XP_016872903.1 | 10 | 1129 | |||
GenBank mRNA | AF144074.1 | - | 11 | 1137 | ||
AJ000332.1 | - | 10 | 1055 | |||
AK026997.1 | - | 10 | 1072 | |||
AK295806.1 | - | 7 | 728 | |||
AK299819.1 | - | 9 | 956 | |||
AK302752.1 | - | 8 | 812 | |||
AK314370.1 | - | 10 | 1071 | |||
BC034439.1 | - | 1 | 58 | |||
BC065266.1 | - | 9 | 949 | 6155944 | ||
D42041.1 | - | 10 | 1053 | |||
EU794613.1 | - | 10 | 1088 |