Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GANAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278192.1 | NP_001265121.1 | 12 | 1525 | ||
NM_001278193.1 | NP_001265122.1 | 11 | 1459 | |||
NM_001278194.1 | NP_001265123.1 | 13 | 1692 | |||
NM_198334.2 | NP_938148.1 | 14 | 1801 | |||
NM_198335.3 | NP_938149.2 | 15 | 1867 | |||
XM_017017412.1 | XP_016872901.1 | 12 | 1535 | |||
XM_017017413.1 | XP_016872902.1 | 14 | 1917 | |||
XM_017017414.1 | XP_016872903.1 | 14 | 1748 | |||
GenBank mRNA | AF144074.1 | - | 15 | 1756 | ||
AJ000332.1 | - | 14 | 1674 | |||
AK026548.1 | - | 3 | 205 | |||
AK026997.1 | - | 14 | 1691 | |||
AK295806.1 | - | 11 | 1347 | |||
AK299819.1 | - | 13 | 1575 | |||
AK302752.1 | - | 12 | 1431 | |||
AK314370.1 | - | 14 | 1690 | |||
BC017433.2 | - | 4 | 306 | |||
BC017435.2 | - | 4 | 306 | |||
BC034439.1 | - | 5 | 677 | |||
BC065266.1 | - | 13 | 1568 | 6155944 | ||
D42041.1 | - | 14 | 1672 | |||
EU794613.1 | - | 14 | 1707 |