Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM120A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286722.1 | NP_001273651.1 | 7, 8 | 1638 | ||
NM_001286723.1 | NP_001273652.1 | 7, 8 | 1641 | |||
NM_001286724.1 | NP_001273653.1 | 7, 8 | 1641 | |||
NM_014612.4 | NP_055427.2 | 7, 8 | 1641 | |||
XM_005251842.4 | XP_005251899.1 | 7, 8 | 1593 | |||
GenBank mRNA | AF214737.1 | - | 7, 8 | 1446 | ||
AF214738.1 | - | 7, 8 | 1446 | |||
BC007879.2 | - | 6, 7 | 953 | |||
BC098584.1 | - | 7, 8 | 1617 | 30531603 | ||
BC111736.1 | - | 7, 8 | 1439 | 40036337 | ||
D80005.2 | - | 7, 8 | 1402 |