Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM120A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286722.1 | NP_001273651.1 | 16 | 3048 | ||
NM_014612.4 | NP_055427.2 | 16 | 3051 | |||
XM_005251842.4 | XP_005251899.1 | 16 | 3000 | |||
XM_011518412.2 | XP_011516714.1 | 17 | 2952 | |||
XM_011518413.2 | XP_011516715.1 | 17 | 2949 | |||
XM_011518414.2 | XP_011516716.1 | 17 | 2949 | |||
GenBank mRNA | AF055017.1 | - | 6 | 959 | ||
AF214737.1 | - | 16 | 2856 | |||
BC098584.1 | - | 16 | 3027 | 30531603 | ||
BC111736.1 | - | 16 | 2849 | 40036337 | ||
D80005.2 | - | 16 | 2812 |