Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM120A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286722.1 | NP_001273651.1 | 18 | 3549 | ||
NM_014612.4 | NP_055427.2 | 18 | 3552 | |||
XM_005251842.4 | XP_005251899.1 | 18 | 3501 | |||
XM_011518412.2 | XP_011516714.1 | 19 | 3453 | |||
XM_011518413.2 | XP_011516715.1 | 19 | 3450 | |||
XM_011518414.2 | XP_011516716.1 | 19 | 3450 | |||
GenBank mRNA | AF055017.1 | - | 8 | 1460 | ||
AK091785.1 | - | 1 | 1634 | |||
AW949787.1 | - | 1 | 267 | |||
AY450393.1 | - | 1 | 1634 | |||
BC098584.1 | - | 18 | 3528 | 30531603 | ||
BC111736.1 | - | 18 | 3350 | 40036337 | ||
D80005.2 | - | 18 | 3313 |