Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSME4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014614.2 | NP_055429.2 | 22, 23 | 2684 | ||
XM_006711969.1 | XP_006712032.1 | 21, 22 | 2261 | |||
XM_011532705.2 | XP_011531007.1 | 23, 24 | 2708 | |||
XM_011532706.2 | XP_011531008.1 | 23, 24 | 2708 | |||
XM_011532707.2 | XP_011531009.1 | 22, 23 | 2642 | |||
XM_011532709.2 | XP_011531011.1 | 23, 24 | 2708 | |||
GenBank mRNA | AB621805.1 | - | 21, 22 | 2801 | ||
AY894754.1 | - | 21, 22 | 2533 | |||
AY894755.1 | - | 3, 4 | 372 | |||
AY894756.1 | - | 22, 23 | 2628 | |||
BC112169.1 | - | 21, 22 | 2320 | 8327637 | ||
BC113668.1 | - | 21, 22 | 2320 | 8322720 | ||
BC143739.1 | - | 21, 22 | 2320 | |||
D38521.1 | - | 22, 23 | 2494 |