Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSME4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014614.2 | NP_055429.2 | 5 | 728 | ||
XM_006711969.1 | XP_006712032.1 | 3 | 244 | |||
XM_011532705.2 | XP_011531007.1 | 6 | 752 | |||
XM_011532706.2 | XP_011531008.1 | 6 | 752 | |||
XM_011532707.2 | XP_011531009.1 | 5 | 686 | |||
XM_011532709.2 | XP_011531011.1 | 6 | 752 | |||
GenBank mRNA | AB621805.1 | - | 4 | 845 | ||
AL599601.1 | - | 5 | 601 | |||
AY894754.1 | - | 5 | 728 | |||
AY894756.1 | - | 5 | 672 | |||
BC112169.1 | - | 4 | 364 | 8327637 | ||
BC113668.1 | - | 4 | 364 | 8322720 | ||
BC143739.1 | - | 4 | 364 | |||
D38521.1 | - | 5 | 538 |