Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOBTB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160036.1 | NP_001153508.1 | 12 | 2416 | ||
NM_001160037.1 | NP_001153509.1 | 10 | 2112 | |||
NM_015178.2 | NP_055993.2 | 10 | 2604 | |||
XM_017013251.1 | XP_016868740.1 | 11 | 6931 | |||
GenBank mRNA | AB018260.2 | - | 10 | 2604 | ||
AK292863.1 | - | 10 | 2252 | |||
AK314366.1 | - | 10 | 2367 | |||
AY009093.1 | - | 10 | 2258 | |||
BC034917.1 | - | 12 | 2416 | 4647716 |