Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145114.1 | NP_001138586.1 | 5 | 748 | ||
NM_001284337.1 | NP_001271266.1 | 4 | 631 | |||
NM_015179.3 | NP_055994.2 | 7 | 931 | |||
XM_011539555.1 | XP_011537857.1 | 6 | 800 | |||
XM_011539556.2 | XP_011537858.1 | 7 | 901 | |||
GenBank mRNA | AB014590.1 | - | 4 | 544 | ||
AK000238.1 | - | 4 | 341 | |||
AK022496.1 | - | 7 | 902 | |||
AK296315.1 | - | 5 | 612 | |||
BC012745.2 | - | 7 | 869 | 4025242 |