Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARHGEF9 SIRNA Assays ›
Gene Symbol:
ARHGEF9
Gene Name:
Cdc42 guanine nucleotide exchange factor 9
Gene Aliases:
COLLYBISTIN, EIEE8, HPEM-2, PEM-2, PEM2
Chromosome Location:
Chr.X: 63634967 - 63786025 on Build GRCh38
UniGene ID:
Hs.54697
Species:
Human
siRNA ID
HSS118405
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARHGEF9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001173479.1
NP_001166950.1
4
820
NM_001173480.1
NP_001166951.1
4
1226
NM_015185.2
NP_056000.1
5
1417
XM_005262249.2
XP_005262306.1
5
1479
XM_005262250.3
XP_005262307.1
5
1377
XM_005262251.1
XP_005262308.1
5
727
XM_005262252.1
XP_005262309.1
6
746
XM_011530890.1
XP_011529192.1
6
1161
XM_011530892.1
XP_011529194.1
5
985
XM_017029363.1
XP_016884852.1
6
1506
XM_017029364.1
XP_016884853.1
7
1283
XM_017029365.1
XP_016884854.1
5
1384
XM_017029366.1
XP_016884855.1
6
1165
XM_017029367.1
XP_016884856.1
6
1154
XM_017029368.1
XP_016884857.1
6
1455
XM_017029369.1
XP_016884858.1
5
1479
XM_017029370.1
XP_016884859.1
5
1388
XM_017029371.1
XP_016884860.1
5
1377
XM_017029372.1
XP_016884861.1
5
727
XM_017029373.1
XP_016884862.1
6
1520
XM_017029374.1
XP_016884863.1
4
1208
XM_017029375.1
XP_016884864.1
5
1385
XM_017029376.1
XP_016884865.1
5
1385
XM_017029377.1
XP_016884866.1
5
1388
XM_017029378.1
XP_016884867.1
5
1479
XM_017029379.1
XP_016884868.1
5
727
XM_017029380.1
XP_016884869.1
4
1208
GenBank mRNA
AB007884.1
-
5
1379
AK289993.1
-
5
831
AK295033.1
-
4
832
AK295178.1
-
5
727
AL832116.1
-
5
988
BC117406.1
-
5
728
40125957
Back To Top
More Information
Panther Classification:
Molecular Function -
guanyl-nucleotide exchange factor
Gene Ontology Categories:
Function(s)
Process(es)
ion transmembrane transport
regulation of Rho protein signal transduction
positive regulation of apoptotic process
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0