Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLCB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015192.3 | NP_056007.1 | 9 | 1156 | ||
NM_182734.2 | NP_877398.1 | 9 | 1156 | |||
XM_011529199.2 | XP_011527501.1 | 9 | 1116 | |||
XM_011529200.2 | XP_011527502.1 | 7 | 780 | |||
XM_011529201.2 | XP_011527503.1 | 7 | 590 | |||
XM_017027752.1 | XP_016883241.1 | 9 | 1116 | |||
XM_017027753.1 | XP_016883242.1 | 9 | 743 | |||
GenBank mRNA | AB011153.3 | - | 9 | 1156 | ||
AJ278313.1 | - | 9 | 772 | |||
AJ278314.1 | - | 9 | 772 | |||
AK127693.1 | - | 7 | 571 | |||
AK299195.1 | - | 9 | 791 | |||
AY004175.1 | - | 9 | 754 | |||
BC069420.1 | - | 9 | 772 | 7262405 | ||
BC117231.1 | - | 9 | 827 | 40125782 |