Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297673.1 | NP_001284602.1 | 11 | 3376 | ||
NM_001297674.1 | NP_001284603.1 | 10 | 3516 | |||
NM_015203.4 | NP_056018.2 | 11 | 3594 | |||
XM_005245033.2 | XP_005245090.1 | 12 | 2655 | |||
XM_006711240.2 | XP_006711303.1 | 12 | 3417 | |||
XM_011509344.2 | XP_011507646.1 | 10 | 3356 | |||
XM_017000763.1 | XP_016856252.1 | 9 | 3278 | |||
XM_017000764.1 | XP_016856253.1 | 11 | 2932 | |||
XM_017000765.1 | XP_016856254.1 | 11 | 3216 | |||
XM_017000766.1 | XP_016856255.1 | 11 | 2289 | |||
XM_017000767.1 | XP_016856256.1 | 10 | 2376 | |||
GenBank mRNA | AB007929.2 | - | 11 | 2963 | ||
AK022984.1 | - | 1 | 750 | |||
AK056707.1 | - | 11 | 3001 | |||
AK125864.1 | - | 12 | 3024 | |||
AK291703.1 | - | 11 | 3012 | |||
AK304380.1 | - | 11 | 2794 | |||
AK308148.1 | - | 10 | 2925 | |||
BC045623.1 | - | 10 | 2917 | 5269891 | ||
BX641025.1 | - | 12 | 3489 |