Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297673.1 | NP_001284602.1 | 11 | 3401 | ||
NM_001297674.1 | NP_001284603.1 | 10 | 3541 | |||
NM_015203.4 | NP_056018.2 | 11 | 3619 | |||
XM_005245033.2 | XP_005245090.1 | 12 | 2680 | |||
XM_006711240.2 | XP_006711303.1 | 12 | 3442 | |||
XM_011509344.2 | XP_011507646.1 | 10 | 3381 | |||
XM_017000763.1 | XP_016856252.1 | 9 | 3303 | |||
XM_017000764.1 | XP_016856253.1 | 11 | 2957 | |||
XM_017000765.1 | XP_016856254.1 | 11 | 3241 | |||
XM_017000766.1 | XP_016856255.1 | 11 | 2314 | |||
XM_017000767.1 | XP_016856256.1 | 10 | 2401 | |||
GenBank mRNA | AB007929.2 | - | 11 | 2988 | ||
AK022984.1 | - | 1 | 775 | |||
AK056707.1 | - | 11 | 3026 | |||
AK125864.1 | - | 12 | 3049 | |||
AK291703.1 | - | 11 | 3037 | |||
AK304380.1 | - | 11 | 2819 | |||
AK308148.1 | - | 10 | 2950 | |||
BC045623.1 | - | 10 | 2942 | 5269891 | ||
BX641025.1 | - | 12 | 3514 |