Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083625.2 | NP_001077094.1 | 12 | 6286 | ||
NM_001204056.1 | NP_001190985.1 | 12 | 6384 | |||
NM_015208.4 | NP_056023.3 | 13 | 6355 | |||
XM_005258092.3 | XP_005258149.1 | 13 | 6488 | |||
XM_005258093.3 | XP_005258150.1 | 13 | 6472 | |||
XM_005258095.2 | XP_005258152.1 | 13 | 6321 | |||
XM_005258096.2 | XP_005258153.1 | 12 | 6256 | |||
XM_011525639.2 | XP_011523941.1 | 14 | 6518 | |||
XM_017025661.1 | XP_016881150.1 | 13 | 6450 | |||
XM_017025662.1 | XP_016881151.1 | 13 | 6495 | |||
XM_017025663.1 | XP_016881152.1 | 12 | 6371 | |||
XM_017025664.1 | XP_016881153.1 | 12 | 6365 | |||
XM_017025665.1 | XP_016881154.1 | 12 | 6399 | |||
XM_017025666.1 | XP_016881155.1 | 12 | 6338 | |||
XM_017025667.1 | XP_016881156.1 | 14 | 6349 | |||
XM_017025668.1 | XP_016881157.1 | 14 | 6343 | |||
XM_017025669.1 | XP_016881158.1 | 7 | 5445 | |||
XM_017025670.1 | XP_016881159.1 | 9 | 5912 | |||
GenBank mRNA | AB020681.1 | - | 5 | 1710 | ||
AF317425.1 | - | 12 | 6091 | |||
AL834204.1 | - | 12 | 6213 | |||
AY373757.1 | - | 13 | 6305 | |||
AY533564.2 | - | 12 | 6384 |