Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017590.5 | NP_060060.3 | 4, 5 | 561 | ||
XM_011530016.2 | XP_011528318.1 | 4, 5 | 542 | |||
GenBank mRNA | AB028954.1 | - | 1, 2 | 174 | ||
AF188530.1 | - | 4, 5 | 389 | |||
AK023849.1 | - | 4, 5 | 349 | |||
AK225891.1 | - | 4, 5 | 534 | |||
AK315009.1 | - | 4, 5 | 503 | |||
BC098378.1 | - | 4, 5 | 499 | |||
BC152558.1 | - | 4, 5 | 539 | 8860405 |