Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLUH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015229.3 | NP_056044.3 | 11 | 2034 | ||
XM_011523768.2 | XP_011522070.1 | 10 | 1962 | |||
GenBank mRNA | AB014564.1 | - | 7 | 1423 | ||
AK023003.1 | - | 3 | 881 | |||
AK125717.1 | - | 11 | 2054 | |||
BC063786.1 | - | 5 | 1113 | |||
CR627449.1 | - | 9 | 1769 |