Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLUH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015229.3 | NP_056044.3 | 14, 15 | 2502 | ||
XM_011523768.2 | XP_011522070.1 | 13, 14 | 2433 | |||
GenBank mRNA | AB014564.1 | - | 10, 11 | 1891 | ||
AK023003.1 | - | 6, 7 | 1352 | |||
AK125717.1 | - | 14, 15 | 2522 | |||
BC063786.1 | - | 8, 9 | 1584 | |||
CR627449.1 | - | 12, 13 | 2240 |