Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SATB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172509.1 | NP_001165980.1 | 11 | 2707 | ||
NM_001172517.1 | NP_001165988.1 | 12 | 2303 | |||
NM_015265.3 | NP_056080.1 | 12 | 2283 | |||
XM_005246396.2 | XP_005246453.1 | 10 | 1938 | |||
XM_006712372.2 | XP_006712435.1 | 11 | 2053 | |||
XM_011510840.2 | XP_011509142.1 | 12 | 2277 | |||
XM_017003656.1 | XP_016859145.1 | 11 | 1938 | |||
GenBank mRNA | AB028957.1 | - | 11 | 1976 | ||
AB209376.1 | - | 12 | 2303 | |||
AK056638.1 | - | 11 | 2708 | |||
AK291463.1 | - | 12 | 2289 | |||
BC098136.1 | - | 10 | 1941 | 40007830 | ||
BC099723.1 | - | 10 | 1941 | 40007832 | ||
BC103492.1 | - | 7 | 1587 | 40007829 | ||
BC103500.1 | - | 10 | 1941 | 40007831 |