Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAN2B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292038.1 | NP_001278967.1 | 9 | 1179 | ||
NM_015274.2 | NP_056089.1 | 9 | 1332 | |||
GenBank mRNA | AB023152.2 | - | 9 | 1291 | ||
AK301473.1 | - | 9 | 1164 | |||
AL832232.1 | - | 9 | 1302 | |||
AL833071.2 | - | 9 | 1305 | |||
BC033307.1 | - | 9 | 1301 | 4932398 | ||
BC094773.1 | - | 9 | 1332 | 30531368 |