Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NEDD4L SIRNA Assays ›
Gene Symbol:
NEDD4L
Gene Name:
neural precursor cell expressed, developmentally down-regulated 4-like, E3 ubiquitin protein ligase
Gene Aliases:
NEDD4-2, NEDD4.2, RSP5, hNEDD4-2
Chromosome Location:
Chr.18: 58044362 - 58401540 on Build GRCh38
UniGene ID:
Hs.185677
Species:
Human
siRNA ID
HSS118599
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NEDD4L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001144964.1
NP_001138436.1
22, 23
2417
NM_001144965.1
NP_001138437.1
22, 23
2369
NM_001144966.2
NP_001138438.1
21, 22
2294
NM_001144967.2
NP_001138439.1
22, 23
2463
NM_001144968.1
NP_001138440.1
22, 23
2359
NM_001144969.1
NP_001138441.1
21, 22
2299
NM_001144970.2
NP_001138442.1
20, 21
2342
NM_001144971.1
NP_001138443.1
22, 23
2387
NM_001243960.1
NP_001230889.1
20, 21
2271
NM_015277.5
NP_056092.2
21, 22
2403
XM_005266658.4
XP_005266715.2
19, 20
2394
XM_005266660.4
XP_005266717.2
18, 19
2338
XM_005266663.4
XP_005266720.2
17, 18
2278
XM_006722421.3
XP_006722484.2
18, 19
2333
XM_006722424.3
XP_006722487.2
15, 16
1886
XM_006722425.3
XP_006722488.2
14, 15
1830
XM_006722426.3
XP_006722489.1
24, 25
2611
XM_006722428.3
XP_006722491.1
23, 24
2533
XM_006722430.3
XP_006722493.1
22, 23
2441
XM_011525887.2
XP_011524189.1
24, 25
2591
XM_017025676.1
XP_016881165.1
16, 17
2083
XM_017025677.1
XP_016881166.1
15, 16
2027
XM_017025678.1
XP_016881167.1
22, 23
2472
XM_017025679.1
XP_016881168.1
21, 22
2387
XM_017025680.1
XP_016881169.1
18, 19
2075
XM_017025681.1
XP_016881170.1
17, 18
1877
GenBank mRNA
AB007899.1
-
20, 21
2242
AB071179.1
-
21, 22
2234
AF210730.1
-
21, 22
2194
AF385931.1
-
22, 23
2418
AK294963.1
-
21, 22
2294
AK300444.1
-
21, 22
2245
AL137469.1
-
17, 18
1715
AY112983.1
-
22, 23
2369
AY112984.2
-
22, 23
2359
AY112985.2
-
21, 22
2303
AY312514.1
-
22, 23
2180
BC000621.2
-
16, 17
1820
BC019345.1
-
16, 17
1820
BC032597.1
-
20, 21
2298
5528964
BQ215431.1
-
1, 2
27
DQ181796.1
-
22, 23
2387
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
protein polyubiquitination
regulation of membrane depolarization
protein monoubiquitination
sodium ion transport
cellular sodium ion homeostasis
excretion
response to metal ion
protein ubiquitination
viral life cycle
water homeostasis
ion transmembrane transport
regulation of ion transmembrane transport
regulation of protein catabolic process
regulation of membrane potential
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of protein catabolic process
positive regulation of endocytosis
regulation of membrane repolarization
protein K48-linked ubiquitination
ventricular cardiac muscle cell action potential
regulation of potassium ion transmembrane transporter activity
negative regulation of potassium ion transmembrane transporter activity
negative regulation of potassium ion transmembrane transport
negative regulation of sodium ion transmembrane transport
positive regulation of dendrite extension
negative regulation of protein localization to cell surface
negative regulation of sodium ion transmembrane transporter activity
positive regulation of caveolin-mediated endocytosis
ubiquitin-protein transferase activity
protein binding
potassium channel regulator activity
ligase activity
sodium channel regulator activity
potassium channel inhibitor activity
sodium channel inhibitor activity
ion channel binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-v5ndd:80/100.66.76.213:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline