Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SASH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015278.3 | NP_056093.3 | 18, 19 | 3812 | ||
XM_005266875.1 | XP_005266932.1 | 11, 12 | 3038 | |||
XM_011535640.1 | XP_011533942.1 | 18, 19 | 3255 | |||
XM_017010598.1 | XP_016866087.1 | 18, 19 | 4533 | |||
XM_017010599.1 | XP_016866088.1 | 18, 19 | 3856 | |||
XM_017010600.1 | XP_016866089.1 | 18, 19 | 3744 | |||
XM_017010601.1 | XP_016866090.1 | 17, 18 | 3958 | |||
XM_017010602.1 | XP_016866091.1 | 16, 17 | 3655 | |||
XM_017010603.1 | XP_016866092.1 | 16, 17 | 3594 | |||
XM_017010604.1 | XP_016866093.1 | 15, 16 | 3503 | |||
XM_017010605.1 | XP_016866094.1 | 15, 16 | 3469 | |||
XM_017010606.1 | XP_016866095.1 | 11, 12 | 3527 | |||
XM_017010607.1 | XP_016866096.1 | 19, 20 | 3435 | |||
GenBank mRNA | AB018333.1 | - | 18, 19 | 3555 | ||
AJ507735.1 | - | 18, 19 | 3343 | |||
AK024403.1 | - | 12, 13 | 2797 | |||
AK025495.1 | - | 18, 19 | 3812 | |||
AK074249.1 | - | 8, 9 | 2097 | |||
BC028303.1 | - | 1, 2 | 323 | |||
BC063279.1 | - | 18, 19 | 3281 | |||
BX537611.1 | - | 7, 8 | 2870 |