Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CLASP1 SIRNA Assays ›
Gene Symbol:
CLASP1
Gene Name:
cytoplasmic linker associated protein 1
Gene Aliases:
MAST1
Chromosome Location:
Chr.2: 121337776 - 121649476 on Build GRCh38
UniGene ID:
Hs.469840
Species:
Human
siRNA ID
HSS118603
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CLASP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142273.1
NP_001135745.1
26, 27
3000
NM_001142274.1
NP_001135746.1
25, 26
2976
NM_001207051.1
NP_001193980.1
25, 26
2994
NM_015282.2
NP_056097.1
25, 26
3060
XM_006712377.1
XP_006712440.1
26, 27
2995
XM_006712381.1
XP_006712444.1
24, 25
2944
XM_006712382.1
XP_006712445.1
24, 25
2920
XM_006712383.1
XP_006712446.1
23, 24
2896
XM_006712385.1
XP_006712448.1
18, 19
1995
XM_011510848.1
XP_011509150.1
25, 26
2947
XM_011510849.1
XP_011509151.1
24, 25
2923
XM_017003657.1
XP_016859146.1
28, 29
3127
XM_017003658.1
XP_016859147.1
28, 29
3127
XM_017003659.1
XP_016859148.1
27, 28
3103
XM_017003660.1
XP_016859149.1
26, 27
2995
XM_017003661.1
XP_016859150.1
27, 28
3079
XM_017003662.1
XP_016859151.1
26, 27
3055
XM_017003663.1
XP_016859152.1
25, 26
2947
XM_017003664.1
XP_016859153.1
24, 25
2944
XM_017003665.1
XP_016859154.1
28, 29
3127
XM_017003666.1
XP_016859155.1
27, 28
3019
XM_017003667.1
XP_016859156.1
27, 28
3079
XM_017003668.1
XP_016859157.1
25, 26
2968
XM_017003669.1
XP_016859158.1
26, 27
3055
XM_017003670.1
XP_016859159.1
25, 26
3028
XM_017003671.1
XP_016859160.1
24, 25
2923
XM_017003672.1
XP_016859161.1
25, 26
2968
XM_017003673.1
XP_016859162.1
26, 27
2968
XM_017003674.1
XP_016859163.1
25, 26
2947
XM_017003675.1
XP_016859164.1
24, 25
2923
XM_017003676.1
XP_016859165.1
21, 22
3961
XM_017003677.1
XP_016859166.1
20, 21
3913
XM_017003678.1
XP_016859167.1
19, 20
3888
XM_017003679.1
XP_016859168.1
19, 20
3886
XM_017003680.1
XP_016859169.1
17, 18
3757
XM_017003681.1
XP_016859170.1
19, 20
3804
XM_017003682.1
XP_016859171.1
19, 20
3801
XM_017003683.1
XP_016859172.1
18, 19
3797
XM_017003684.1
XP_016859173.1
20, 21
2894
XM_017003685.1
XP_016859174.1
18, 19
3779
XM_017003686.1
XP_016859175.1
17, 18
3776
XM_017003687.1
XP_016859176.1
19, 20
2870
XM_017003688.1
XP_016859177.1
17, 18
3755
XM_017003689.1
XP_016859178.1
19, 20
2785
GenBank mRNA
AB014522.1
-
17, 18
1923
AF347693.1
-
24, 25
2835
AK074338.1
-
11, 12
1234
AK091140.1
-
24, 25
2934
AK294922.1
-
2, 3
287
AK297259.1
-
19, 20
1951
AK299331.1
-
18, 19
2480
AK302393.1
-
18, 19
2012
BC112940.1
-
25, 26
2920
5501463
BC132723.1
-
26, 27
2962
40146745
BC144107.1
-
25, 26
2956
9052627
BX648933.1
-
7, 8
709
CR933722.1
-
18, 19
1928
Back To Top
More Information
Panther Classification:
Molecular Function -
non-motor microtubule binding protein
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
microtubule cytoskeleton organization
microtubule bundle formation
vesicle targeting
microtubule nucleation
negative regulation of microtubule depolymerization
Golgi organization
mitotic spindle organization
sister chromatid cohesion
establishment or maintenance of cell polarity
exit from mitosis
regulation of gastrulation
positive regulation of epithelial cell migration
regulation of epithelial to mesenchymal transition
astral microtubule organization
microtubule organizing center organization
negative regulation of microtubule polymerization or depolymerization
positive regulation of microtubule polymerization
microtubule anchoring
establishment of mitotic spindle localization
positive regulation of exocytosis
establishment of spindle orientation
cell division
negative regulation of stress fiber assembly
regulation of focal adhesion assembly
regulation of microtubule cytoskeleton organization
positive regulation of extracellular matrix disassembly
establishment of epithelial cell polarity
mitotic spindle assembly
negative regulation of wound healing, spreading of epidermal cells
positive regulation of basement membrane assembly involved in embryonic body morphogenesis
dystroglycan binding
protein binding
microtubule binding
kinetochore binding
microtubule plus-end binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-9xzrh:80/100.66.79.6:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline