Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CLASP1 SIRNA Assays ›
Gene Symbol:
CLASP1
Gene Name:
cytoplasmic linker associated protein 1
Gene Aliases:
MAST1
Chromosome Location:
Chr.2: 121337776 - 121649476 on Build GRCh38
UniGene ID:
Hs.469840
Species:
Human
siRNA ID
HSS118604
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CLASP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142273.1
NP_001135745.1
34
3811
NM_001142274.1
NP_001135746.1
33
3787
NM_001207051.1
NP_001193980.1
33
3805
NM_015282.2
NP_056097.1
34
3988
XM_006712377.1
XP_006712440.1
34
3806
XM_006712381.1
XP_006712444.1
32
3755
XM_006712382.1
XP_006712445.1
32
3731
XM_006712383.1
XP_006712446.1
31
3707
XM_006712385.1
XP_006712448.1
27
2875
XM_011510848.1
XP_011509150.1
34
3827
XM_011510849.1
XP_011509151.1
33
3803
XM_017003657.1
XP_016859146.1
38
4109
XM_017003658.1
XP_016859147.1
36
3938
XM_017003659.1
XP_016859148.1
35
3914
XM_017003660.1
XP_016859149.1
35
3908
XM_017003661.1
XP_016859150.1
35
3890
XM_017003662.1
XP_016859151.1
34
3866
XM_017003663.1
XP_016859152.1
34
3860
XM_017003664.1
XP_016859153.1
33
3857
XM_017003665.1
XP_016859154.1
36
3938
XM_017003666.1
XP_016859155.1
35
3830
XM_017003667.1
XP_016859156.1
35
3890
XM_017003668.1
XP_016859157.1
33
3779
XM_017003669.1
XP_016859158.1
34
3866
XM_017003670.1
XP_016859159.1
33
3839
XM_017003671.1
XP_016859160.1
32
3734
XM_017003672.1
XP_016859161.1
33
3779
XM_017003673.1
XP_016859162.1
34
3779
XM_017003674.1
XP_016859163.1
33
3758
XM_017003675.1
XP_016859164.1
32
3734
XM_017003676.1
XP_016859165.1
29
4772
XM_017003677.1
XP_016859166.1
28
4724
XM_017003678.1
XP_016859167.1
27
4699
XM_017003679.1
XP_016859168.1
27
4697
XM_017003680.1
XP_016859169.1
25
4568
XM_017003681.1
XP_016859170.1
27
4615
XM_017003682.1
XP_016859171.1
27
4612
XM_017003683.1
XP_016859172.1
26
4608
XM_017003684.1
XP_016859173.1
28
3705
XM_017003685.1
XP_016859174.1
26
4590
XM_017003686.1
XP_016859175.1
25
4587
XM_017003687.1
XP_016859176.1
27
3681
XM_017003688.1
XP_016859177.1
25
4566
XM_017003689.1
XP_016859178.1
27
3596
GenBank mRNA
AB014522.1
-
26
2851
AF347693.1
-
33
3763
AK074338.1
-
19
2045
AK123216.1
-
1
1098
AK294922.1
-
10
1098
AK297259.1
-
27
2762
AK297420.1
-
3
310
AK299331.1
-
26
3291
AK301814.1
-
5
548
AK302393.1
-
27
2892
BC112940.1
-
33
3731
5501463
BC132723.1
-
34
3773
40146745
BC144107.1
-
33
3767
9052627
BX648523.1
-
3
528
BX648933.1
-
16
1637
CR749627.1
-
6
669
CR933722.1
-
27
2808
Back To Top
More Information
Panther Classification:
Molecular Function -
non-motor microtubule binding protein
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
microtubule cytoskeleton organization
microtubule bundle formation
vesicle targeting
microtubule nucleation
negative regulation of microtubule depolymerization
Golgi organization
mitotic spindle organization
sister chromatid cohesion
establishment or maintenance of cell polarity
exit from mitosis
regulation of gastrulation
positive regulation of epithelial cell migration
regulation of epithelial to mesenchymal transition
astral microtubule organization
microtubule organizing center organization
negative regulation of microtubule polymerization or depolymerization
positive regulation of microtubule polymerization
microtubule anchoring
establishment of mitotic spindle localization
positive regulation of exocytosis
establishment of spindle orientation
cell division
negative regulation of stress fiber assembly
regulation of focal adhesion assembly
regulation of microtubule cytoskeleton organization
positive regulation of extracellular matrix disassembly
establishment of epithelial cell polarity
mitotic spindle assembly
negative regulation of wound healing, spreading of epidermal cells
positive regulation of basement membrane assembly involved in embryonic body morphogenesis
dystroglycan binding
protein binding
microtubule binding
kinetochore binding
microtubule plus-end binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0