Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CLASP1 SIRNA Assays ›
Gene Symbol:
CLASP1
Gene Name:
cytoplasmic linker associated protein 1
Gene Aliases:
MAST1
Chromosome Location:
Chr.2: 121337776 - 121649476 on Build GRCh38
UniGene ID:
Hs.469840
Species:
Human
siRNA ID
HSS118605
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CLASP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142273.1
NP_001135745.1
39, 40
4746
NM_001142274.1
NP_001135746.1
38, 39
4722
NM_001207051.1
NP_001193980.1
38, 39
4740
NM_015282.2
NP_056097.1
39, 40
4923
XM_006712377.1
XP_006712440.1
39, 40
4741
XM_006712381.1
XP_006712444.1
37, 38
4690
XM_006712382.1
XP_006712445.1
37, 38
4666
XM_006712383.1
XP_006712446.1
36, 37
4642
XM_006712385.1
XP_006712448.1
32, 33
3810
XM_011510848.1
XP_011509150.1
39, 40
4762
XM_011510849.1
XP_011509151.1
38, 39
4738
XM_017003657.1
XP_016859146.1
43, 44
5044
XM_017003665.1
XP_016859154.1
41, 42
4873
XM_017003667.1
XP_016859156.1
40, 41
4825
XM_017003669.1
XP_016859158.1
39, 40
4801
XM_017003670.1
XP_016859159.1
38, 39
4774
XM_017003672.1
XP_016859161.1
38, 39
4714
XM_017003673.1
XP_016859162.1
39, 40
4714
XM_017003674.1
XP_016859163.1
38, 39
4693
XM_017003675.1
XP_016859164.1
37, 38
4669
XM_017003676.1
XP_016859165.1
34, 35
5707
XM_017003677.1
XP_016859166.1
33, 34
5659
XM_017003678.1
XP_016859167.1
32, 33
5634
XM_017003679.1
XP_016859168.1
32, 33
5632
XM_017003681.1
XP_016859170.1
32, 33
5550
XM_017003682.1
XP_016859171.1
32, 33
5547
XM_017003683.1
XP_016859172.1
31, 32
5543
XM_017003684.1
XP_016859173.1
33, 34
4640
XM_017003685.1
XP_016859174.1
31, 32
5525
XM_017003686.1
XP_016859175.1
30, 31
5522
XM_017003687.1
XP_016859176.1
32, 33
4616
XM_017003688.1
XP_016859177.1
30, 31
5501
XM_017003689.1
XP_016859178.1
32, 33
4531
GenBank mRNA
AB014522.1
-
31, 32
3786
AF347693.1
-
38, 39
4698
AK074338.1
-
25, 26
2981
AK123216.1
-
6, 7
2033
AK294922.1
-
15, 16
2033
AK297259.1
-
32, 33
3697
AK297420.1
-
8, 9
1245
AK299331.1
-
31, 32
4226
AK301814.1
-
10, 11
1483
AK302393.1
-
32, 33
3827
BC032563.1
-
4, 5
552
BC112940.1
-
38, 39
4666
5501463
BC132723.1
-
39, 40
4708
40146745
BC144107.1
-
38, 39
4702
9052627
BX648933.1
-
21, 22
2572
CR749627.1
-
11, 12
1603
CR933722.1
-
32, 33
3743
Back To Top
More Information
Panther Classification:
Molecular Function -
non-motor microtubule binding protein
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
microtubule cytoskeleton organization
microtubule bundle formation
vesicle targeting
microtubule nucleation
negative regulation of microtubule depolymerization
Golgi organization
mitotic spindle organization
sister chromatid cohesion
establishment or maintenance of cell polarity
exit from mitosis
regulation of gastrulation
positive regulation of epithelial cell migration
regulation of epithelial to mesenchymal transition
astral microtubule organization
microtubule organizing center organization
negative regulation of microtubule polymerization or depolymerization
positive regulation of microtubule polymerization
microtubule anchoring
establishment of mitotic spindle localization
positive regulation of exocytosis
establishment of spindle orientation
cell division
negative regulation of stress fiber assembly
regulation of focal adhesion assembly
regulation of microtubule cytoskeleton organization
positive regulation of extracellular matrix disassembly
establishment of epithelial cell polarity
mitotic spindle assembly
negative regulation of wound healing, spreading of epidermal cells
positive regulation of basement membrane assembly involved in embryonic body morphogenesis
dystroglycan binding
protein binding
microtubule binding
kinetochore binding
microtubule plus-end binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-cp9cp:80/100.66.77.247:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline