Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015285.2 | NP_056100.2 | 27 | 4437 | ||
NM_052834.2 | NP_443066.2 | 26 | 4338 | |||
XM_006722431.2 | XP_006722494.1 | 27 | 4369 | |||
XM_017025682.1 | XP_016881171.1 | 26 | 4243 | |||
XM_017025683.1 | XP_016881172.1 | 17 | 2830 | |||
GenBank mRNA | AB011113.2 | - | 27 | 4400 | ||
AF188125.1 | - | 25 | 4127 | |||
AY099325.1 | - | 26 | 4226 | |||
BC050352.1 | - | 13 | 1598 | |||
BC131558.1 | - | 15 | 2201 | 40108655 | ||
BC150282.1 | - | 27 | 4400 | 9007268 |