Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287811.1 | NP_001274740.1 | 7 | 1015 | ||
NM_015291.3 | NP_056106.1 | 8 | 1200 | |||
NR_109898.1 | 8 | 1200 | ||||
GenBank mRNA | AB023179.2 | - | 8 | 1156 | ||
AK023805.1 | - | 4 | 370 | |||
AK091848.1 | - | 8 | 1158 | |||
AK292479.1 | - | 8 | 1132 | |||
AL833299.1 | - | 8 | 1139 | |||
BC047363.1 | - | 8 | 1200 | 5269539 | ||
BX648982.1 | - | 8 | 1125 | |||
CR749259.1 | - | 7 | 960 | |||
CR749567.1 | - | 7 | 960 |